2-(Trifluoromethyl)-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC400181-1G
PC400181-250MG
APOSPC400181-1GEA
0
GBP
APOSPC400181-1G
APOSPC400181-250MG
2-(Trifluoromethyl)-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
2-(Trifluoromethyl)-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula:
C₁₃H₁₇BF₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18726737 CAS Number: 1612287-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...