Nonafluoropentanoic anhydride 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
PC32845-100G
PC32845-50G
APOSPC32845-100GEA
279
GBP
APOSPC32845-100G
APOSPC32845-50G
Nonafluoropentanoic anhydride 97%
Nonafluoropentanoic anhydride
Formula:
C₁₀F₁₈O₃ MW: 510.076 g/mol Boiling Pt: 137 °C Storage Temperature: Ambient |
MDL Number:
MFCD06248659 CAS Number: 308-28-1 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...