2-Fluoroterephthalonitrile 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC201186-1G PC201186-250MG PC201186-5G
APOSPC201186-1GEA 92.3 GBP
APOSPC201186-1G APOSPC201186-250MG APOSPC201186-5G
2-Fluoroterephthalonitrile 98%
2-Fluoroterephthalonitrile
Formula: C₈H₃FN₂
Storage Temperature: Ambient
MDL Number: MFCD01657004
CAS Number: 1897-53-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR