Cadmium(II) tetrafluoroborate 50% in aqueous solution

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC1625-100G PC1625-500G
APOSPC1625-100GEA 300 GBP
APOSPC1625-100G APOSPC1625-500G
Cadmium(II) tetrafluoroborate 50% in aqueous solution
Cadmium(II) tetrafluoroborate
Formula: Cd(BF₄)₂
Storage Temperature: Ambient
MDL Number: MFCD00015993
CAS Number: 14486-19-2
UN: 2570
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR