2,3,5,6-Tetrafluoronitrobenzene 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2,4,5-Tetrafluoro-3-nitrobenzene

PC10437-1G PC10437-5G PC10437-250MG
APOSPC10437-1GEA 27.5 GBP
APOSPC10437-1G APOSPC10437-5G APOSPC10437-250MG
2,3,5,6-Tetrafluoronitrobenzene 97%
2,3,5,6-Tetrafluoronitrobenzene
Formula: C₆HF₄NO₂
Boiling Pt: 93…94 °C (52 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02093333
CAS Number: 6257-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR