2',6'-Dichloro-4'-fluoroacetanilide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC10432-100G PC10432-25G
APOSPC10432-100GEA 302 GBP
APOSPC10432-100G APOSPC10432-25G
2',6'-Dichloro-4'-fluoroacetanilide
2',6'-Dichloro-4'-fluoroacetanilide
Formula: C₈H₆Cl₂FNO
Melting Pt: 186…187 °C
Storage Temperature: Ambient
MDL Number: MFCD07368695
CAS Number: 392-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR