1-Benzylpyrazole 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR959890-10G
OR959890-1G
OR959890-5G
APOSOR959890-10GEA
415
GBP
APOSOR959890-10G
APOSOR959890-1G
APOSOR959890-5G
1-Benzylpyrazole 98%
1-Benzylpyrazole
Formula:
C₁₀H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00462231 CAS Number: 10199-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...