Ethyl 2-chloroethylcarbamate 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR953267-1G
OR953267-5G
APOSOR953267-1GEA
111
GBP
APOSOR953267-1G
APOSOR953267-5G
Ethyl 2-chloroethylcarbamate 95%
Ethyl 2-chloroethylcarbamate
Formula:
C₅H₁₀ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04038117 CAS Number: 6329-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...