3-Cyano-N-ethylbenzamide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR946462-1G OR946462-5G
APOSOR946462-1GEA 158 GBP
APOSOR946462-1G APOSOR946462-5G
3-Cyano-N-ethylbenzamide 98%
3-Cyano-N-ethylbenzamide
Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09938849
CAS Number: 623569-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR