3-(Chloromethyl)-N-ethylbenzamide 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR946372-10G
OR946372-1G
OR946372-5G
APOSOR946372-10GEA
363
GBP
APOSOR946372-10G
APOSOR946372-1G
APOSOR946372-5G
3-(Chloromethyl)-N-ethylbenzamide 98%
3-(Chloromethyl)-N-ethylbenzamide
Formula:
C₁₀H₁₂ClNO MW: 197.661 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11208367 CAS Number: 1094431-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...