2-Amino-1-(2-bromophenyl)ethanol hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR943727-1G OR943727-500MG
APOSOR943727-1GEA 483 GBP
APOSOR943727-1G APOSOR943727-500MG
2-Amino-1-(2-bromophenyl)ethanol hydrochloride 97%
2-Amino-1-(2-bromophenyl)ethanol hydrochloride
Formula: C₈H₁₀BrNO·ClH
Storage Temperature: Ambient
MDL Number: MFCD05861635
CAS Number: 71095-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR