N-Methylcyclobutanamine hydrochloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR941585-1G OR941585-250MG
APOSOR941585-1GEA 109 GBP
APOSOR941585-1G APOSOR941585-250MG
N-Methylcyclobutanamine hydrochloride 95%
N-Methylcyclobutanamine hydrochloride
Formula: C₅H₁₁N·ClH
Storage Temperature: Ambient
MDL Number: MFCD11506003
CAS Number: 848497-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR