N-Phenylcyclobutanecarboxamide 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR941229-1G
OR941229-5G
APOSOR941229-1GEA
237
GBP
APOSOR941229-1G
APOSOR941229-5G
N-Phenylcyclobutanecarboxamide 95%
N-Phenylcyclobutanecarboxamide
Formula:
C₁₁H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD03784889 CAS Number: 15907-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...