1-Hydroxymethylbenzocyclobutene 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bicyclo[4.2.0]octa-1,3,5-trien-7-ylmethanol

OR931311-1G OR931311-250MG
APOSOR931311-1GEA 110 GBP
APOSOR931311-1G APOSOR931311-250MG
1-Hydroxymethylbenzocyclobutene 95%
1-Hydroxymethylbenzocyclobutene
Formula: C₉H₈O
MW: 132.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09752655
CAS Number: 15100-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR