Ethyl 2-amino-3-cyclobutylpropanoate 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR930722-1G
OR930722-250MG
APOSOR930722-1GEA
352
GBP
APOSOR930722-1G
APOSOR930722-250MG
Ethyl 2-amino-3-cyclobutylpropanoate 95%
Ethyl 2-amino-3-cyclobutylpropanoate
Formula:
C₉H₁₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11111478 CAS Number: 394735-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...