Methyl 2-[methyl(pyridin-2-yl)amino]acetate 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR930052-1G
OR930052-250MG
APOSOR930052-1GEA
237
GBP
APOSOR930052-1G
APOSOR930052-250MG
Methyl 2-[methyl(pyridin-2-yl)amino]acetate 95%
Methyl 2-[methyl(pyridin-2-yl)amino]acetate
Formula:
C₉H₁₂N₂O₂ MW: 180.204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16151670 CAS Number: 1250236-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...