Ethyl 3-hydroxy-2-phenylacrylate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR929736-1G OR929736-250MG
APOSOR929736-1GEA 315 GBP
APOSOR929736-1G APOSOR929736-250MG
Ethyl 3-hydroxy-2-phenylacrylate 95%
Ethyl 3-hydroxy-2-phenylacrylate
Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD24842546
CAS Number: 76636-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR