tert-Butyl 3-oxocyclopentylcarbamate 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
tert-Butyl (3-oxocyclopentyl)carbamate
OR928719-10G
OR928719-1G
OR928719-5G
APOSOR928719-10GEA
286
GBP
APOSOR928719-10G
APOSOR928719-1G
APOSOR928719-5G
tert-Butyl 3-oxocyclopentylcarbamate 95%
tert-Butyl 3-oxocyclopentylcarbamate
Formula:
C₁₀H₁₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09751875 CAS Number: 847416-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...