α-methyl-2-furanethanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR927143-1G OR927143-250MG
APOSOR927143-1GEA 315 GBP
APOSOR927143-1G APOSOR927143-250MG
α-methyl-2-furanethanol 95%
α-methyl-2-furanethanol
Formula: C₇H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD15145897
CAS Number: 66040-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR