3-Bromoazetidine hydrochloride 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR923770-1G
OR923770-250MG
APOSOR923770-1GEA
51.5
GBP
APOSOR923770-1G
APOSOR923770-250MG
3-Bromoazetidine hydrochloride 95%
3-Bromoazetidine hydrochloride
Formula:
C₃H₆BrN·ClH Storage Temperature: Ambient |
MDL Number:
MFCD09834130 CAS Number: 53913-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...