2-Cyano-6-isopropylpyridine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR923642-1G OR923642-250MG
APOSOR923642-1GEA 583 GBP
APOSOR923642-1G APOSOR923642-250MG
2-Cyano-6-isopropylpyridine 95%
2-Cyano-6-isopropylpyridine
Formula: C₉H₁₀N₂
MW: 146.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384527
CAS Number: 337904-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR