2,4-Pyrimidinedicarbonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR923029-1G OR923029-250MG
APOSOR923029-1GEA 583 GBP
APOSOR923029-1G APOSOR923029-250MG
2,4-Pyrimidinedicarbonitrile 95%
2,4-Pyrimidinedicarbonitrile
Formula: C₆H₂N₄
Storage Temperature: Ambient
MDL Number: MFCD09834972
CAS Number: 75928-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR