Trideuteriomethyl 4-methylbenzenesulphonate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methy-d3 (toluenesulphonate), Methyl-D3p-toluenesulphonate

OR909752-1G OR909752-5G
APOSOR909752-1GEA 21.2 GBP
APOSOR909752-1G APOSOR909752-5G
Trideuteriomethyl 4-methylbenzenesulphonate 98%
Trideuteriomethyl 4-methylbenzenesulphonate
Formula: C₈H₇D₃O₃S
MW: 189.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069406
CAS Number: 7575-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR