2-Amino-5-iodo-3-nitropyridine 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Iodo-3-nitropyridin-2-amine

OR8989-10G OR8989-5G
APOSOR8989-10GEA 120 GBP
APOSOR8989-10G APOSOR8989-5G
2-Amino-5-iodo-3-nitropyridine 95+%
2-Amino-5-iodo-3-nitropyridine
Formula: C₅H₄IN₃O₂
MW: 265.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038464
CAS Number: 25391-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR