1-Bromo-8-phenyloctane 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR8184-1G OR8184-5G
APOSOR8184-1GEA 45.2 GBP
APOSOR8184-1G APOSOR8184-5G
1-Bromo-8-phenyloctane 95+%
1-Bromo-8-phenyloctane
Formula: C₁₄H₂₁Br
Boiling Pt: 84…86 °C (0.15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01075178
CAS Number: 54646-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR