N-Boc-N,N'-dimethylethylamine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: tert-Butyl methyl(2-(methylamino)ethyl)carbamate

OR55058-1G OR55058-5G
APOSOR55058-1GEA 15.9 GBP
APOSOR55058-1G APOSOR55058-5G
N-Boc-N,N'-dimethylethylamine 97%
N-Boc-N,N'-dimethylethylamine
Formula: C₉H₂₀N₂O₂
MW: 188.267 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09953267
CAS Number: 112257-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR