3-Hydroxy-4-methoxypyridine-2-carboxylic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Hydroxy-4-methoxypicolinic acid

OR54549-1G OR54549-250MG
APOSOR54549-1GEA 179 GBP
APOSOR54549-1G APOSOR54549-250MG
3-Hydroxy-4-methoxypyridine-2-carboxylic acid 97%
3-Hydroxy-4-methoxypyridine-2-carboxylic acid
Formula: C₇H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD18254272
CAS Number: 210300-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR