3H-Benzotriazole-4-carboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR54545-1G OR54545-250MG
APOSOR54545-1GEA 890 GBP
APOSOR54545-1G APOSOR54545-250MG
3H-Benzotriazole-4-carboxylic acid
3H-Benzotriazole-4-carboxylic acid
Formula: C₇H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00869366
CAS Number: 62972-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR