5-Bromo-2-methylpyridine-N-oxide 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR54544-1G OR54544-5G
APOSOR54544-1GEA 127 GBP
APOSOR54544-1G APOSOR54544-5G
5-Bromo-2-methylpyridine-N-oxide 95%
5-Bromo-2-methylpyridine-N-oxide
Formula: C₆H₆BrNO
MW: 188.022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00234172
CAS Number: 31181-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR