3,3'-Bis(allyloxy)-[1,1'-biphenyl]-4,4'-diamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(4-Amino-3-prop-2-enoxyphenyl)-2-prop-2-enoxyaniline

OR52963-10G OR52963-1G OR52963-5G
APOSOR52963-10GEA 108 GBP
APOSOR52963-10G APOSOR52963-1G APOSOR52963-5G
3,3'-Bis(allyloxy)-[1,1'-biphenyl]-4,4'-diamine
3,3'-Bis(allyloxy)-[1,1'-biphenyl]-4,4'-diamine
Formula: C₁₈H₂₀N₂O₂
MW: 296.364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32708567
CAS Number: 2573217-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR