N,N-Dimethyl-4-(tributylstannyl)aniline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Tributylstannyl)-1-dimethylaminobenzene, [4-(Dimethylamino)phenyl]tributylstannane

OR52021-1G OR52021-250MG
APOSOR52021-1GEA 233 GBP
APOSOR52021-1G APOSOR52021-250MG
N,N-Dimethyl-4-(tributylstannyl)aniline
N,N-Dimethyl-4-(tributylstannyl)aniline
Formula: C₂₀H₃₇NSn
Storage Temperature: Refrigerator
MDL Number: MFCD20481826
CAS Number: 84600-70-4
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR