4-(Tributylstannyl)anisole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (4-Methoxyphenyl)tributylstannane, 1-Methoxy-4-(tributylstannyl)benzene

OR52017-1G OR52017-5G
APOSOR52017-1GEA 158 GBP
APOSOR52017-1G APOSOR52017-5G
4-(Tributylstannyl)anisole 98%
4-(Tributylstannyl)anisole
Formula: C₁₉H₃₄OSn
Storage Temperature: Ambient
MDL Number: MFCD01114690
CAS Number: 70744-47-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR