α-(3-Methoxyphenyl)-3-methyloxybenzylamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR470477-1G OR470477-5G
APOSOR470477-1GEA 324 GBP
APOSOR470477-1G APOSOR470477-5G
α-(3-Methoxyphenyl)-3-methyloxybenzylamine
α-(3-Methoxyphenyl)-3-methyloxybenzylamine
Formula: C₁₅H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD12605374
CAS Number: 860598-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR