Ethyl (S)-1-Phenylethylcarbamate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR470088-25G OR470088-5G
APOSOR470088-25GEA 553 GBP
APOSOR470088-25G APOSOR470088-5G
Ethyl (S)-1-Phenylethylcarbamate
Ethyl (S)-1-Phenylethylcarbamate
Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00137647
CAS Number: 33290-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR