3,3',5,5'-Tetraisopropyldiphenoquinone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR460027-1G OR460027-250MG
APOSOR460027-1GEA 300 GBP
APOSOR460027-1G APOSOR460027-250MG
3,3',5,5'-Tetraisopropyldiphenoquinone
3,3',5,5'-Tetraisopropyldiphenoquinone
Formula: C₂₄H₃₂O₂
MW: 352.51 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Ambient
MDL Number: MFCD18072169
CAS Number: 2178-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR