1-(3-Methoxypyridin-2-yl)piperazine hydrochloride (1:1)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR460026-1G OR460026-250MG
APOSOR460026-1GEA 286 GBP
APOSOR460026-1G APOSOR460026-250MG
1-(3-Methoxypyridin-2-yl)piperazine hydrochloride (1:1)
1-(3-Methoxypyridin-2-yl)piperazine hydrochloride (1:1)
Formula: C₁₀H₁₅N₃O·ClH
MW: 229.707 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074398
CAS Number: 1260082-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR