4-(Tributylstannyl)-1,2-dimethylbenzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR399039-1G OR399039-250MG
APOSOR399039-1GEA 231 GBP
APOSOR399039-1G APOSOR399039-250MG
4-(Tributylstannyl)-1,2-dimethylbenzene
4-(Tributylstannyl)-1,2-dimethylbenzene
Formula: C₂₀H₃₆Sn
Storage Temperature: Ambient
CAS Number: 1505287-81-9
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR