1-(Tributylstannyl)-3,5-bistrifluoromethylbenzene
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR399038-1G
OR399038-250MG
APOSOR399038-1GEA
255
GBP
APOSOR399038-1G
APOSOR399038-250MG
1-(Tributylstannyl)-3,5-bistrifluoromethylbenzene
1-(Tributylstannyl)-3,5-bistrifluoromethylbenzene
Formula:
C₂₀H₃₀F₆Sn Storage Temperature: Ambient |
CAS Number:
717918-62-2 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...