3-(Tributylstannyl)-1-chlorobenzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tributyl-(3-chlorophenyl)stannane

OR399037-1G OR399037-250MG
APOSOR399037-1GEA 231 GBP
APOSOR399037-1G APOSOR399037-250MG
3-(Tributylstannyl)-1-chlorobenzene
3-(Tributylstannyl)-1-chlorobenzene
Formula: C₁₈H₃₁ClSn
MW: 401.602 g/mol
Storage Temperature: Ambient
CAS Number: 24344-58-9
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR