4-(Tributylstannyl)-1,2-methylenedioxybenzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR399036-1G OR399036-250MG
APOSOR399036-1GEA 231 GBP
APOSOR399036-1G APOSOR399036-250MG
4-(Tributylstannyl)-1,2-methylenedioxybenzene
4-(Tributylstannyl)-1,2-methylenedioxybenzene
Formula: C₁₉H₃₂O₂Sn
Storage Temperature: Ambient
CAS Number: 124010-49-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR