4-(Tributylstannyl)-1-phenoxybenzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tributyl-(4-phenoxyphenyl)stannane

OR399034-1G OR399034-250MG
APOSOR399034-1GEA 231 GBP
APOSOR399034-1G APOSOR399034-250MG
4-(Tributylstannyl)-1-phenoxybenzene
4-(Tributylstannyl)-1-phenoxybenzene
Formula: C₂₄H₃₆OSn
MW: 459.252 g/mol
Storage Temperature: Ambient
CAS Number: 99128-02-6
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR