4-(Tributylstannyl)-1-ethylbenzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR399032-1G OR399032-250MG
APOSOR399032-1GEA 231 GBP
APOSOR399032-1G APOSOR399032-250MG
4-(Tributylstannyl)-1-ethylbenzene
4-(Tributylstannyl)-1-ethylbenzene
Formula: C₂₀H₃₆Sn
Storage Temperature: Ambient
CAS Number: 198224-53-2
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR