(1S,2S,5R,6S,7S)-6,8,8-Trimethyl-3-azatricyclo[5.1.1.02,5]nonan-4-one

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR307673-1G OR307673-250MG
APOSOR307673-1GEA 856 GBP
APOSOR307673-1G APOSOR307673-250MG
(1S,2S,5R,6S,7S)-6,8,8-Trimethyl-3-azatricyclo[5.1.1.02,5]nonan-4-one
(1S,2S,5R,6S,7S)-6,8,8-Trimethyl-3-azatricyclo[5.1.1.02,5]nonan-4-one
Formula: C₁₁H₁₇NO
MW: 179.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505102
CAS Number: 1000304-40-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR