N,N-Dimethyloctanamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: N-Octanoyldimethylamine, N,N-Dimethylcaprylamide, Octanoic acid dimethylamide

OR307187-25G OR307187-5G
APOSOR307187-25GEA 111 GBP
APOSOR307187-25G APOSOR307187-5G
N,N-Dimethyloctanamide
N,N-Dimethyloctanamide
Formula: C₁₀H₂₁NO
MW: 171.28 g/mol
Boiling Pt: 187 °C (100 mm Hg)
Melting Pt: –21 °C
Density: 0.881 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00043763
CAS Number: 1118-92-9
EINECS: 214-272-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR