4-(Tributylstannyl)-1H-indole, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (1H-Indol-4-yl)tributylstannane

OR15624-1G OR15624-250MG
APOSOR15624-1GEA 147 GBP
APOSOR15624-1G APOSOR15624-250MG
4-(Tributylstannyl)-1H-indole, tech
4-(Tributylstannyl)-1H-indole
Formula: C₂₀H₃₃NSn
Storage Temperature: Refrigerator
MDL Number: MFCD09265481
CAS Number: 1025745-98-5
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR