Cadmium(II) nitrate hydrate 99,999%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN1354-250G IN1354-50G
APOSIN1354-250GEA 263 GBP
APOSIN1354-250G APOSIN1354-50G
Cadmium(II) nitrate hydrate 99,999%
Cadmium(II) nitrate hydrate
Formula: Cd·(NO₃)₂·H₂O
Boiling Pt: 132 °C
Melting Pt: 59.4 °C
Storage Temperature: Ambient
MDL Number: MFCD32066826
CAS Number: 79990-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR