Fluorinert™ FC-72

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC3312I-100G PC3312I-25G PC3312I-500G
APOSPC3312I-100GEA 47.5 GBP
APOSPC3312I-100G APOSPC3312I-25G APOSPC3312I-500G
Fluorinert™ FC-72
Fluorinert™

Caution: Fluorinert is a trademark of 3M Company

Boiling Pt: 58…60 °C
Melting Pt: –4 °C
Density: 1.669 g/cm3 @ 25°C
Storage Temperature: Ambient
MDL Number: MFCD00000437
CAS Number: 86508-42-1
EINECS: 206-585-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR