Tris(2-hydroxyethyl)-1,3,5-triazinetrione, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,3,5-Tris(2-hydroxyethyl)isocyanuric acid, 1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione, 1,3,5-Tris(2-hydroxyethyl)isocyanurate
M-Clarity™ quality level = MQ100
309001-250G
309001-50G
SIAL309001-250GEA
449
GBP
SIAL309001-250G
SIAL309001-50G
Tris(2-hydroxyethyl)-1,3,5-triazinetrione, Sigma-Aldrich®
Tris(2-hydroxyethyl)-1,3,5-triazinetrione
Formula:
C₉H₁₅N₃O₆ MW: 261.23 g/mol Boiling Pt: 527 °C (1013 hPa) Melting Pt: 133…136 °C |
MDL Number:
MFCD00003549 CAS Number: 839-90-7 EINECS: 212-660-9 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...