2-Chloro-4-nitropyridine ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
L17699.03
L17699.06
L17699.03EA
51.7
GBP
L17699.03
L17699.06
2-Chloro-4-nitropyridine ≥98%
2-Chloro-4-nitropyridine
Purity: 98%
CAS No.: 23056-36-2
Molecular Formula: C5H3ClN2O2
Formula Weight: 158.54
Melting Point: 50-54°
Beilstein Reference: 120418
Formula:
C₅H₃ClN₂O₂ Melting Pt: 50…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00661454 CAS Number: 23056-36-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...