2-Acetylpyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002741-100G 002741-25G
FLUO002741-100GEA 0 GBP
FLUO002741-100G FLUO002741-25G
2-Acetylpyrazine
2-Acetylpyrazine
Formula: C₆H₆N₂O
Boiling Pt: 59 °C (33 mmHg)
Melting Pt: 79…80 °C
MDL Number: MFCD00006134
CAS Number: 22047-25-2
EINECS: 244-753-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR